N,N-Diethyl-m-toluamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diethyl-m-toluamide, Diethyltoluamide, N,N-Diethyl-3-methylbenzamide, Deet
A17486-0E
A17486-22
A17486-36
CAAAA17486-0EEA
385.52
CAD
CAAAA17486-0E
CAAAA17486-22
CAAAA17486-36
N,N-Diethyl-m-toluamide 97%
N,N-Diethyl-m-toluamide
Purity: 97%
CAS No.: 134-62-3
Molecular Formula: C12H17NO
Formula Weight: 191.27
Flash Point: 145°(293°F)
Refractive Index: 1.5230
Boiling Point: 110-112°/1mm
Beilstein Reference: 2046711
Merck Reference: 14,2856
MDL No.: MFCD00009046
Formula:
C₁₂H₁₇NO Boiling Pt: 110…112 °C/1 mm Melting Pt: –45 °C Density: 0.997 Flash Pt: 145 °C(293 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009046 CAS Number: 134-62-3 EINECS: 205-149-7 Merck Index: 14,2856 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...