1-Naphthyl acetate 99%

Supplier: Thermo Scientific Chemicals

A17482-18 A17482-30
CAAAA17482-18EA 87.53 CAD
CAAAA17482-18 CAAAA17482-30
1-Naphthyl acetate 99%
1-Naphthyl acetate

Purity: 99% CAS No.: 830-81-9 Molecular Formula: C12H10O2 Formula Weight: 186.21 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 43-46° Beilstein Reference: 2046403 MDL No.: MFCD00003922

Formula: C₁₂H₁₀O₂
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 43…46 °C
Density: 1.1305 g/cm³ (50 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00003922
CAS Number: 830-81-9
EINECS: 212-599-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR