1-(2-Pyridylazo)-2-naphthol (PAN) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: PAN, 1-(2-Pyridylazo)-2-naphthol

A17475-03 A17475-06 A17475-14
CAAAAA17475-03EA 51.67 CAD
CAAAAA17475-03 CAAAA17475-06 CAAAA17475-14
1-(2-Pyridylazo)-2-naphthol (PAN) 98%
1-(2-Pyridylazo)-2-naphthol

Purity: 98% CAS No.: 85-85-8 Molecular Formula: C15H11N3O Formula Weight: 249.27 Melting Point: 138-141° Beilstein Reference: 206390

Formula: C₁₅H₁₁N₃O
MW: 249.27 g/mol
Boiling Pt: 412 °C (1013 hPa)
Melting Pt: 138…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00004071
CAS Number: 85-85-8
EINECS: 201-637-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR