2-Fluorophenyl isocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17466-06
A17466-14
CAAAA17466-06EA
71.26
CAD
CAAAA17466-06
CAAAA17466-14
2-Fluorophenyl isocyanate 98%
2-Fluorophenyl isocyanate
Purity: 98%
CAS No.: 16744-98-2
Molecular Formula: C7H4FNO
Formula Weight: 137.11
Storage Temperature: 2°C to 8°C
Flash Point: 50°(122°F)
Refractive Index: 1.5125
Odor: Pungent, makes eyes water
Boiling Point: 56-58°/10mm
Beilstein Reference: 1210127
UN No.: 3080
MDL No.: MFCD00001996
Formula:
C₇H₄FNO Boiling Pt: 56…58 °C/10 mm Density: 1.221 Flash Pt: 50 °C (122 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001996 CAS Number: 16744-98-2 EINECS: 240-804-0 UN: 3080 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...