Sodium propionate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17440-0E
A17440-36
CAAAA17440-0EEA
186.44
CAD
CAAAA17440-0E
CAAAA17440-36
Sodium propionate 99%
Sodium propionate
Purity: 99%
CAS No.: 137-40-6
Molecular Formula: C3H5NaO2
Formula Weight: 96.06
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 285-291°
Merck Reference: 14,8669
MDL No.: MFCD00002759
Formula:
C₃H₅NaO₂ Melting Pt: 285…291 °C Flash Pt: 157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002759 CAS Number: 137-40-6 EINECS: 205-290-4 Merck Index: 14,8669 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...