α-Bromo-2,6-difluorotoluene 97+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(Bromomethyl)-1,3-difluorobenzene, 2,6-Difluorobenzyl bromide

A17434-06 A17434-14
CAAAA17434-06EA 92.75 CAD
CAAAA17434-06 CAAAA17434-14
α-Bromo-2,6-difluorotoluene 97+%
α-Bromo-2,6-difluorotoluene

Purity: 97+% CAS No.: 85118-00-9 Molecular Formula: C7H5BrF2 Formula Weight: 207.02 Flash Point: 110°(230°F) Melting Point: 49-53° Beilstein Reference: 2083943 UN No.: 1759 MDL No.: MFCD00000329

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Melting Pt: 49…53 °C
Density: 1.600
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000329
CAS Number: 85118-00-9
EINECS: 285-652-6
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR