Tetrachlorophthalic anhydride 98%

Supplier: Thermo Scientific Chemicals
Danger

A17404-0C A17404-0E A17404-36
CAAAA17404-0CEA 878.48 CAD
CAAAA17404-0C CAAAA17404-0E CAAAA17404-36
Tetrachlorophthalic anhydride 98%
Tetrachlorophthalic anhydride

Purity: 98% CAS No.: 117-08-8 Molecular Formula: C8Cl4O3 Formula Weight: 285.90 Flash Point: 362°(683°F) Odor: Weak Boiling Point: 349-354° Beilstein Reference: 211560 UN No.: 3077 MDL No.: MFCD00005920

Formula: C₈Cl₄O₃
MW: 285.9 g/mol
Boiling Pt: 371
Melting Pt: 254…258 °C
Flash Pt: 362 °C (683 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005920
CAS Number: 117-08-8
EINECS: 204-171-4
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR