1,3-Dibromotetrafluorobenzene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17392-06
A17392-14
CAAAA17392-03EA
146.29
CAD
CAAAA17392-03
CAAAA17392-06
CAAAA17392-14
1,3-Dibromotetrafluorobenzene 98+%
1,3-Dibromotetrafluorobenzene
Purity: 98+%
CAS No.: 1559-87-1
Molecular Formula: C6Br2F4
Formula Weight: 307.87
Flash Point: 92°(198°F)
Refractive Index: 1.5160
Boiling Point: 85-88°/18mm
MDL No.: MFCD00031533
Formula:
C₆Br₂F₄ Boiling Pt: 85…88 °C/18 mm Density: 2,180 Flash Pt: 92 °C(198 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00031533 CAS Number: 1559-87-1 EINECS: 216-326-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...