Eosin B

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Eosin bluish, 2,7-Dinitro-4,5-dibromofluorescein, Eosin B disodium salt, Sodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate, 2-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt, Acid Red 91

A17377-14 A17377-22
CAAAA17377-14EA 73.63 CAD
CAAAA17377-14 CAAAA17377-22
Eosin B
Eosin B

A counterstain for collagen


CAS No.: 548-24-3 Molecular Formula: C20H6Br2N2Na2O9 Formula Weight: 624.08 MDL No.: MFCD00005041

Formula: C₂₀H₆Br₂N₂Na₂O₉
MW: 624.06 g/mol
Melting Pt: > 310 °C
Storage Temperature: Ambient
MDL Number: MFCD00005041
CAS Number: 548-24-3
EINECS: 208-943-1
Merck Index: 12,03638

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR