Triisopropylchlorosilane 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17376-30
A17376-09
A17376-18
CAAAAA17376-30EA
1741.16
CAD
CAAAAA17376-30
CAAAA17376-09
CAAAA17376-18
Triisopropylchlorosilane 97+%
Triisopropylchlorosilane
Purity: 97+%
CAS No.: 13154-24-0
Molecular Formula: [(CH3)2CH]3SiCl
Formula Weight: 192.81
Flash Point: 62°(144°F)
Refractive Index: 1.4535
Odor: Acrid
Boiling Point: 200°
Beilstein Reference: 1737446
UN No.: 2987
MDL No.: MFCD00009656
Formula:
C₉H₂₁ClSi Boiling Pt: 200 °C Density: 0.908 Flash Pt: 62 °C (144 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009656 CAS Number: 13154-24-0 UN: 2987 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...