tert-Butyl perbenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17373-14
A17373-22
A17373-36
CAAAAA17373-14EA
45.98
CAD
CAAAAA17373-14
CAAAA17373-22
CAAAA17373-36
tert-Butyl perbenzoate 98%
tert-Butyl perbenzoate
Purity: 98%
CAS No.: 614-45-9
Molecular Formula: C11H14O3
Formula Weight: 194.23
Storage Temperature: 2°C to 8°C
Flash Point: 93°(199°F)
Refractive Index: 1.4990
Odor: Sweet
Melting Point: 6-8°
Boiling Point: 75-76°/0.2mm
Beilstein Reference: 1342734
UN No.: 3103
MDL No.: MFCD00008802
Formula:
C₁₁H₁₄O₃ Boiling Pt: 75…76 °C/0.2 mm Melting Pt: 6…8 °C Density: 1.042 Flash Pt: 93 °C(199 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008802 CAS Number: 614-45-9 EINECS: 210-382-2 UN: 3103 ADR: "5.2", |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...