N,N-Dimethylhexylamine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17364-06
A17364-14
A17364-22
CAAAA17364-06EA
78.68
CAD
CAAAA17364-06
CAAAA17364-14
CAAAA17364-22
N,N-Dimethylhexylamine 99%
N,N-Dimethylhexylamine
Purity: 99%
CAS No.: 4385-04-0
Molecular Formula: CH3(CH2)5N(CH3)2
Formula Weight: 129.25
Flash Point: 33°(92°F)
Refractive Index: 1.4136
Boiling Point: 146-150°
UN No.: 2733
MDL No.: MFCD00009522
Formula:
C₈H₁₉N Boiling Pt: 146…150 °C Density: 0.744 Flash Pt: 33 °C (92 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009522 CAS Number: 4385-04-0 EINECS: 224-491-8 UN: 2734 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...