8-Aminocaprylic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: ω-Aminooctoic acid, 8-Aminooctanoic acid, ω-Aminocaprylic acid, ω-Aminooctanoic acid, 8-Aminooctoic acid

A17353-03 A17353-06
CAAAA17353-03EA 99.38 CAD
CAAAA17353-03 CAAAA17353-06
8-Aminocaprylic acid 99%
8-Aminocaprylic acid

Purity: 99% CAS No.: 1002-57-9 Molecular Formula: NH2(CH2)7CO2H Formula Weight: 159.23 Melting Point: 193-196° MDL No.: MFCD00008245

Formula: C₈H₁₇NO₂
MW: 159.23 g/mol
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: 193…196 °C
Density: 1.221 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008245
CAS Number: 1002-57-9
EINECS: 213-687-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR