2-Ethylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17352-22
A17352-36
CAAAA17352-22EA
273.34
CAD
CAAAA17352-22
CAAAA17352-36
2-Ethylpyridine 98%
2-Ethylpyridine
Purity: 98%
CAS No.: 100-71-0
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 37°(98°F)
Refractive Index: 1.4970
Odor: Unpleasant
pH: 9-10
Melting Point: -63°
Boiling Point: 149°
Beilstein Reference: 106480
UN No.: 1993
MDL No.: MFCD00006361
Formula:
C₇H₉N MW: 107.16 g/mol Boiling Pt: 149 °C Melting Pt: –63 °C Density: 0.937 Flash Pt: 37 °C (98 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006361 CAS Number: 100-71-0 EINECS: 202-881-9 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...