Bis(4-hydroxyphenyl) sulfone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxyphenyl sulfone, 4,4'-Sulphonyldiphenol, 4,4'-Dihydroxy diphenyl sulphone, 4,4'-Dihydroxy diphenyl sulfone, 4,4'-sulfonyldiphenol, 4,4'-Sulfonyldiphenol, 4-Hydroxyphenyl sulphone, 4,4'-sulphonyldiphenol

A17342-0B A17342-30
CAAAA17342-0BEA 260.7 CAD
CAAAA17342-0B CAAAA17342-30
Bis(4-hydroxyphenyl) sulfone 99%
Bis(4-hydroxyphenyl) sulphone

Purity: 99% CAS No.: 80-09-1 Molecular Formula: C12H10O4S Formula Weight: 250.28 Melting Point: 245-250° Beilstein Reference: 2052954

Formula: C₁₂H₁₀O₄S
MW: 250.28 g/mol
Melting Pt: 245…250 °C
Density: 1.366
Storage Temperature: Ambient
MDL Number: MFCD00002350
CAS Number: 80-09-1
EINECS: 201-250-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR