2-Phenylcyclohexanone 98%

Supplier: Thermo Scientific Chemicals

A17340-03 A17340-06 A17340-14
CAAAA17340-03EA 83.42 CAD
CAAAA17340-03 CAAAA17340-06 CAAAA17340-14
2-Phenylcyclohexanone 98%
2-Phenylcyclohexanone

Purity: 98% CAS No.: 1444-65-1 Molecular Formula: C12H14O Formula Weight: 174.24 Melting Point: 56-59° MDL No.: MFCD00001629

Formula: C₁₂H₁₄O
Boiling Pt: 90 °C (0.5 -rr)
Melting Pt: 56…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00001629
CAS Number: 1444-65-1
EINECS: 215-888-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR