Methyl trifluoroacetate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17338-0B
A17338-18
A17338-30
CAAAA17338-0BEA
1600.54
CAD
CAAAA17338-0B
CAAAA17338-18
CAAAA17338-30
Methyl trifluoroacetate 98+%
Methyl trifluoroacetate
Purity: 98+%
CAS No.: 431-47-0
Molecular Formula: CF3CO2CH3
Formula Weight: 128.05
Flash Point: -18°(-0.4°F)
Refractive Index: 1.2900
Odor: Pungent
Melting Point: -78°
Boiling Point: 43-44°
Beilstein Reference: 1756070
UN No.: 2924
MDL No.: MFCD00000417
Formula:
C₃H₃F₃O₂ Boiling Pt: 43…44 °C Melting Pt: –78 °C Density: 1.283 Flash Pt: –18 °C (–0.4 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000417 CAS Number: 431-47-0 EINECS: 207-074-5 UN: 2924 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...