Triethylammonium chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Triethylamine hydrochloride

A17318-0B A17318-30
CAAAA17318-0BEA 230.68 CAD
CAAAA17318-0B CAAAA17318-30
Triethylammonium chloride 98%
Triethylammonium chloride

Protein purification reagent


Purity: 98% CAS No.: 554-68-7 Molecular Formula: C6H15N.HCl Formula Weight: 137.65 Odor: Amine-like pH: 5 Melting Point: ca 260° dec. Beilstein Reference: 3906409 MDL No.: MFCD00012500

Formula: C₆H₁₆ClN
MW: 137.65 g/mol
Melting Pt: ∼ 260 °C (dec.)
Density: 1.07
Storage Temperature: Ambient
MDL Number: MFCD00012500
CAS Number: 554-68-7
EINECS: 209-067-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR