4-Fluoro-2,6-dinitrophenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17309-03
A17309-06
CAAAA17309-03EA
68.57
CAD
CAAAA17309-03
CAAAA17309-06
4-Fluoro-2,6-dinitrophenol 98%
4-Fluoro-2,6-dinitrophenol
Purity: 98%
CAS No.: 364-32-9
Molecular Formula: C6H3FN2O5
Formula Weight: 202.10
Melting Point: 50-56°
MDL No.: MFCD00179333
Formula:
C₆H₃FN₂O₅ MW: 202.1 g/mol Melting Pt: 50…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00179333 CAS Number: 364-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...