1,5-Diiodopentane 97% stabilized

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with copper

A17298-14 A17298-22
CAAAA17298-14EA 88.16 CAD
CAAAA17298-14 CAAAA17298-22
1,5-Diiodopentane 97% stabilized
1,5-Diiodopentane

Purity: 97% CAS No.: 628-77-3 Molecular Formula: I(CH2)5I Formula Weight: 323.94 Flash Point: 110°(230°F) Refractive Index: 1.6012 Boiling Point: 128-130°/10mm Beilstein Reference: 1697281 MDL No.: MFCD00001101

Formula: C₅H₁₀I₂
Boiling Pt: 128…130 °C/10 mm
Melting Pt: 9 °C
Density: 2.176
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001101
CAS Number: 628-77-3
EINECS: 211-054-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR