2-Methylbenzimidazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methyl-1H-benzo[d]imidazole, 2-Methyl-1H-benzimidazole

A17290-18 A17290-30
CAAAA17290-18EA 68.41 CAD
CAAAA17290-18 CAAAA17290-30
2-Methylbenzimidazole 98%
2-Methylbenzimidazole

Purity: 98% CAS No.: 615-15-6 Molecular Formula: C8H8N2 Formula Weight: 132.17 Melting Point: 174-178° Beilstein Reference: 112264

Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 339
Melting Pt: 174…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00005598
CAS Number: 615-15-6
EINECS: 210-411-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR