2-Chloroethyl isocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Isocyanic acid 2-Chloroethyl ester
A17289-22
A17289-14
CAAAAA17289-22EA
455.04
CAD
CAAAAA17289-22
CAAAA17289-14
2-Chloroethyl isocyanate 97%
2-Chloroethyl isocyanate
Purity: 97%
CAS No.: 1943-83-5
Molecular Formula: ClCH2CH2NCO
Formula Weight: 105.52
Storage Temperature: 2°C to 8°C
Flash Point: 56°(132°F)
Refractive Index: 1.4470
Odor: Pungent
Boiling Point: 135°
Beilstein Reference: 1071429
UN No.: 3080
MDL No.: MFCD00002043
Formula:
C₃H₄ClNO MW: 105.52 g/mol Boiling Pt: 135 °C Density: 1.237 Flash Pt: 56 °C (132 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002043 CAS Number: 1943-83-5 EINECS: 217-734-4 UN: 3080 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...