1-Decanol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17288-0F
A17288-AP
CAAAA17288-0FEA
160.99
CAD
CAAAA17288-0F
CAAAA17288-AP
1-Decanol 98+%
1-Decanol
MDL: MFCD00004747
Beilstein: 1735221
Soluble in alcohol, ether. Insoluble in water
Purity: 98+%
CAS No.: 112-30-1
Molecular Formula: C10H22O
Formula Weight: 158.29
Flash Point: 82°(179°F)
Physical Form: Liquid
Density: 0.828
Refractive Index: 1.4370
Appearance: Colorless
Odor: Sweet
Melting Point: 6°
Boiling Point: 230-232°
Solubility: Soluble in alcohol, ether. Insoluble in water
Beilstein Reference: 1735221
Merck Reference: 14,2855
UN No.: 3082
MDL No.: MFCD00004747
Formula:
H₃C(CH₂)₈CH₂OH Boiling Pt: 230…232 °C Melting Pt: 6 °C Density: 0.828 Flash Pt: 82 °C(179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004747 CAS Number: 112-30-1 EINECS: 203-956-9 UN: 3082 ADR: 9,III Merck Index: 14,2855 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...