2,6-Di-tert-butylphenol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Di(dimethylethyl)-2-hydroxybenzene

A17244-0I A17244-0B A17244-30
CAAAAA17244-0IEA 290.72 CAD
CAAAAA17244-0I CAAAA17244-0B CAAAA17244-30
2,6-Di-tert-butylphenol 98%
2,6-Di-tert-butylphenol

Purity: 98% CAS No.: 128-39-2 Molecular Formula: C14H22O Formula Weight: 206.33 Flash Point: 120°(248°F) Odor: Phenol-like Boiling Point: 252-253° Beilstein Reference: 1841887 UN No.: 3077 MDL No.: MFCD00008820

Formula: C₁₄H₂₂O
MW: 206.33 g/mol
Boiling Pt: 252…253 °C
Melting Pt: 34…39 °C
Density: 0.914
Flash Pt: 120 °C (248 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008820
CAS Number: 128-39-2
EINECS: 204-884-0
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR