α,α'-Dibromo-o-xylene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2-Bis(bromomethyl)benzene, o-Xylylene bromide, o-Xylylene dibromide, alpha,alpha'-Dibromo-o-xylene, o-Xylene dibromide

A17241-06 A17241-14 A17241-22
CAAAA17241-06EA 43.43 CAD
CAAAA17241-06 CAAAA17241-14 CAAAA17241-22
α,α'-Dibromo-o-xylene 97%
α,α'-Dibromo-o-xylene

Purity: 97% CAS No.: 91-13-4 Molecular Formula: C8H8Br2 Formula Weight: 263.96 Melting Point: 92-96° Beilstein Reference: 637159 UN No.: 3261 MDL No.: MFCD00000175

Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 140
Melting Pt: 92…96 °C
Density: 1.960
Storage Temperature: Ambient
MDL Number: MFCD00000175
CAS Number: 91-13-4
EINECS: 202-042-7
UN: 2928
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR