3-Aminophenylboronic acid hemisulfate 98+%

Supplier: Thermo Scientific Chemicals
Warning

A17240-03 A17240-06 A17240-14
CAAAA17240-03EA 59.41 CAD
CAAAA17240-03 CAAAA17240-06 CAAAA17240-14
3-Aminophenylboronic acid hemisulfate 98+%
3-Aminophenylboronic acid hemisulphate

Purity: 98+% CAS No.: 66472-86-4 Molecular Formula: C6H8BNO2.0.5H2SO4 Formula Weight: 185.98 Melting Point: 300° Beilstein Reference: 8094151

Formula: C₁₂H₁₈B₂N₂O₈S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00013111
CAS Number: 66472-86-4
EINECS: 266-376-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR