4-Decylbenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

A17234-14 A17234-06
CAAAAA17234-14EA 913.24 CAD
CAAAAA17234-14 CAAAA17234-03 CAAAA17234-06
4-Decylbenzoyl chloride 98%
4-Decylbenzoyl chloride

Purity: 98% CAS No.: 54256-43-8 Molecular Formula: C17H25ClO Formula Weight: 280.84 Flash Point: 110°(230°F) Boiling Point: 171-173°/2mm UN No.: 3265 MDL No.: MFCD00041724

Formula: C₁₇H₂₅ClO
Boiling Pt: 171…173 °C/2 mm
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041724
CAS Number: 54256-43-8
EINECS: 259-046-7
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR