Isopropyl formate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17222-18
A17222-30
CAAAA17222-18EA
103.65
CAD
CAAAA17222-18
CAAAA17222-30
Isopropyl formate 98%
Purity: 98%
CAS No.: 625-55-8
Molecular Formula: C4H8O2
Formula Weight: 88.11
Flash Point: -6°(21°F)
Refractive Index: 1.3685
Melting Point: -93°
Boiling Point: 68-71°
Beilstein Reference: 1735844
UN No.: 1281
MDL No.: MFCD00014127
Formula:
C₄H₈O₂ Boiling Pt: 68…71 °C Melting Pt: –93 °C Density: 0.880 Flash Pt: –6 °C (21 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014127 CAS Number: 625-55-8 EINECS: 210-901-2 UN: 1281 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...