3,3'-Thiodipropionic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,3-Thiodipropionic acid

A17220-0B A17220-30
CAAAA17220-0BEA 129.88 CAD
CAAAA17220-0B CAAAA17220-30
3,3'-Thiodipropionic acid 98%
3,3'-Thiodipropionic acid

Purity: 98% CAS No.: 111-17-1 Molecular Formula: C6H10O4S Formula Weight: 178.21 Melting Point: 127-134° Beilstein Reference: 1210299 Merck Reference: 14,9332 MDL No.: MFCD00002781

Formula: C₆H₁₀O₄S
MW: 178.21 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 127…134 °C
Density: 1.496 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002781
CAS Number: 111-17-1
EINECS: 203-841-3
Merck Index: 14,9332

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR