1-(2-Hydroxyethyl)piperazine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-(2-Hydroxyethyl)piperazine, 2-(1-Piperazinyl)ethanol, 2-Piperazin-1-ylethanol
A17215-09
A17215-22
A17215-36
CAAAAA17215-09EA
48.51
CAD
CAAAAA17215-09
CAAAA17215-22
CAAAA17215-36
1-(2-Hydroxyethyl)piperazine 98+%
1-(2-Hydroxyethyl)piperazine
Purity: 98+%
CAS No.: 103-76-4
Molecular Formula: C6H14N2O
Formula Weight: 130.19
Flash Point: 135°(275°F)
Refractive Index: 1.5065
Odor: Amine-like
pH: 12.3
Melting Point: -39°
Boiling Point: 242-246°
Beilstein Reference: 104361
MDL No.: MFCD00005970
Formula:
C₆H₁₄N₂O Boiling Pt: 242…246 °C Melting Pt: –39 °C Density: 1.060 Flash Pt: 135 °C(275 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005970 CAS Number: 103-76-4 EINECS: 203-142-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...