MOPS-Na 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-(N-Morpholino)propanesulphonic acid sodium salt, Sodium 3-morpholinopropane-1-sulphonate, Sodium 4-morpholin-1-ylpropylsulphonate, 3-(N-Morpholino)propanesulfonic acid sodium salt, Sodium 3-morpholinopropane-1-sulfonate, MOPS sodium salt, Sodium 4-morpholin-1-ylpropylsulfonate
A17214-14
A17214-22
CAAAA17214-14EA
68.26
CAD
CAAAA17214-14
CAAAA17214-22
MOPS-Na 98%
MOPS-Na
Purity: 98%
CAS No.: 71119-22-7
Molecular Formula: C7H14NNaO4S
Formula Weight: 231.25
Physical Form: Cryst powder
Appearance: White
Melting Point: ca 260° dec.
MDL No.: MFCD00064350
Formula:
C₇H₁₄NNaO₄S MW: 231.25 g/mol Melting Pt: ∼ 260 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064350 CAS Number: 71119-22-7 EINECS: 428-420-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...