4-(Trifluoromethoxy)aniline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17208-06
A17208-14
CAAAA17208-06EA
51.98
CAD
CAAAA17208-06
CAAAA17208-14
4-(Trifluoromethoxy)aniline 98%
4-(Trifluoromethoxy)aniline
Purity: 98%
CAS No.: 461-82-5
Molecular Formula: C7H6F3NO
Formula Weight: 177.13
Flash Point: 81°(178°F)
Refractive Index: 1.4630
Odor: Characteristic
Boiling Point: 73-75°/10mm
Beilstein Reference: 2090209
UN No.: 2810
MDL No.: MFCD00041314
Formula:
C₇H₆F₃NO MW: 177.13 Boiling Pt: 73 Density: 1.310 Flash Pt: 81 °C (178 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041314 CAS Number: 461-82-5 EINECS: 207-317-5 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...