(±)-2-Bromopentane tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17205-22
A17205-36
CAAAA17205-22EA
391.84
CAD
CAAAA17205-22
CAAAA17205-36
(±)-2-Bromopentane tech. 90%, Technical Grade
(±)-2-Bromopentane
Purity: 90%
CAS No.: 107-81-3
Molecular Formula: C5H11Br
Formula Weight: 151.05
Flash Point: 20°(68°F)
Refractive Index: 1.4403
Odor: Characteristic
Melting Point: -96°
Boiling Point: 117-118°
Beilstein Reference: 1718829
UN No.: 2343
MDL No.: MFCD00000160
Formula:
H₃CBrCH(CH₂)₂CH₃ Boiling Pt: 117…118 °C Melting Pt: –96 °C Density: 1.220 Flash Pt: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000160 CAS Number: 107-81-3 EINECS: 203-521-3 UN: 2343 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...