1-Naphthyl isocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17200-06
A17200-14
CAAAA17200-06EA
90.06
CAD
CAAAA17200-06
CAAAA17200-14
1-Naphthyl isocyanate 98%
1-Naphthyl isocyanate
Purity: 98%
CAS No.: 86-84-0
Molecular Formula: C11H7NO
Formula Weight: 169.18
Storage Temperature: 2°C to 8°C
Flash Point: 135°(275°F)
Refractive Index: 1.6335
Odor: Irritating
Melting Point: 4°
Boiling Point: 269°
Beilstein Reference: 742779
Merck Reference: 14,6410
UN No.: 2206
MDL No.: MFCD00003881
Formula:
C₁₁H₇NO Boiling Pt: 269 °C Melting Pt: 4 °C Density: 1.177 Flash Pt: 135 °C (275 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003881 CAS Number: 86-84-0 EINECS: 201-703-7 UN: 2206 ADR: 6.1,III Merck Index: 14,6410 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...