2,6-Dibromo-p-toluidine 98+%

Supplier: Thermo Scientific Chemicals
Warning

A17184-14 A17184-22
CAAAA17184-14EA 95.59 CAD
CAAAA17184-14 CAAAA17184-22
2,6-Dibromo-p-toluidine 98+%
2,6-Dibromo-p-toluidine

Purity: 98+% CAS No.: 6968-24-7 Molecular Formula: C7H7Br2N Formula Weight: 264.96 Melting Point: 72-76° Beilstein Reference: 2085782

Formula: C₇H₇Br₂N
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 72…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00007641
CAS Number: 6968-24-7
EINECS: 230-182-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR