α-Chloro-p-xylene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17183-14
A17183-30
CAAAAA17183-14EA
38.71
CAD
CAAAAA17183-14
CAAAA17183-30
α-Chloro-p-xylene 98%
α-Chloro-p-xylene
Purity: 98%
CAS No.: 104-82-5
Molecular Formula: C8H9Cl
Formula Weight: 140.61
Flash Point: 75°(167°F)
Refractive Index: 1.5330
Odor: Pungent
Melting Point: 4-6°
Boiling Point: 199-201°
Beilstein Reference: 507387
Merck Reference: 14,10091
UN No.: 3265
MDL No.: MFCD00000919
Formula:
C₈H₉Cl Boiling Pt: 199…201 °C Melting Pt: 4…6 °C Density: 1.064 Flash Pt: 75 °C (167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000919 CAS Number: 104-82-5 EINECS: 203-241-1 UN: 3265 ADR: 8,III Merck Index: 14,10091 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...