1,4-Di-tert-butylbenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

A17146-06 A17146-14
CAAAA17146-06EA 102.7 CAD
CAAAA17146-06 CAAAA17146-14
1,4-Di-tert-butylbenzene 98%
1,4-Di-tert-butylbenzene

Purity: 98% CAS No.: 1012-72-2 Molecular Formula: C14H22 Formula Weight: 190.33 Melting Point: 75-79° Boiling Point: 236° MDL No.: MFCD00008836

Formula: C₁₄H₂₂
Boiling Pt: 236 °C
Melting Pt: 75…79 °C
Density: 0.985
Storage Temperature: Ambient
MDL Number: MFCD00008836
CAS Number: 1012-72-2
EINECS: 213-790-9
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR