1,3-Diethyl-2-thiourea 98%

Supplier: Thermo Scientific Chemicals
Danger

A17143-0B A17143-30
CAAAA17143-0BEA 180.12 CAD
CAAAA17143-0B CAAAA17143-30
1,3-Diethyl-2-thiourea 98%

Purity: 98% CAS No.: 105-55-5 Molecular Formula: (CH3CH2NH)2C=S Formula Weight: 132.23 Flash Point: 186°(367°F) Melting Point: 75-79° Beilstein Reference: 773905 MDL No.: MFCD00004925

Formula: C₅H₁₂N₂S
Boiling Pt: 169 °C (1013 hPa)
Melting Pt: 75…79 °C
Density: 1.120
Flash Pt: 186 °C(367 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004925
CAS Number: 105-55-5
EINECS: 203-308-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR