2-Adamantylamine hydrochloride 98+%

Supplier: Thermo Scientific Chemicals
Warning

A17140-03 A17140-06 A17140-14
CAAAAA17140-03EA 47.72 CAD
CAAAAA17140-03 CAAAA17140-06 CAAAA17140-14
2-Adamantylamine hydrochloride 98+%
2-Adamantylamine hydrochloride

Purity: 98+% CAS No.: 10523-68-9 Molecular Formula: C10H17N.HCl Formula Weight: 187.71 Melting Point: 300° Beilstein Reference: 4297901

Formula: C₁₀H₁₈ClN
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00074743
CAS Number: 10523-68-9
EINECS: 234-074-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR