Dichlorodibutylgermane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17138-06
A17138-14
CAAAA17138-06EA
442.4
CAD
CAAAA17138-06
CAAAA17138-14
Dichlorodibutylgermane 97%
Dichlorodibutylgermane
Purity: 97%
CAS No.: 4593-81-1
Molecular Formula: C8H18Cl2Ge
Formula Weight: 257.73
Flash Point: 107°(225°F)
Refractive Index: 1.4730
Boiling Point: 241-243°
UN No.: 2922
MDL No.: MFCD00013590
Formula:
C₈H₁₈Cl₂Ge Boiling Pt: 241…243 °C Density: 1.208 Flash Pt: 107 °C (225 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013590 CAS Number: 4593-81-1 EINECS: 224-986-9 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...