Triacetin (glycerol triacetate) 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Glyceryl triacetate, Propane-1,2,3-triyl triacetate, Glycerol triacetate

A17132-0E A17132-36
CAAAA17132-0EEA 189.6 CAD
CAAAA17132-0E CAAAA17132-36
Triacetin (glycerol triacetate) 99%
Triacetin (Glycerol triacetate)

Purity: 99% CAS No.: 102-76-1 Molecular Formula: C9H14O6 Formula Weight: 218.21 Flash Point: 149°(300°F) Physical Form: Liquid Density: 1.155 Odor: Petroleum-like Melting Point: 2-4° Boiling Point: 257-259° MDL No.: MFCD00008716

Formula: C₉H₁₄O₆
Boiling Pt: 257…259 °C
Melting Pt: 2…4 °C
Density: 1.155
Flash Pt: 149 °C(300 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008716
CAS Number: 102-76-1
EINECS: 203-051-9
Merck Index: 13,09664

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR