N,N'-Diethylethylenediamine 96%

Supplier: Thermo Scientific Chemicals
Danger

A17113-14 A17113-06 A17113-22
CAAAAA17113-14EA 287.56 CAD
CAAAAA17113-14 CAAAA17113-06 CAAAA17113-22
N,N'-Diethylethylenediamine 96%
N,N'-Diethylethylenediamine

Purity: 96% CAS No.: 111-74-0 Molecular Formula: CH3CH2NHCH2CH2NHCH2CH3 Formula Weight: 116.21 Flash Point: 33°(92°F) Refractive Index: 1.4326 Boiling Point: 152-154° UN No.: 2685 MDL No.: MFCD00009033

Formula: C₆H₁₆N₂
Boiling Pt: 152…154 °C
Melting Pt: –11 °C
Density: 0.811
Flash Pt: 33 °C (92 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009033
CAS Number: 111-74-0
EINECS: 203-902-4
UN: 2685
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR