3-Bromo-5-iodobenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17076-22
A17076-06
A17076-14
CAAAAA17076-22EA
1216.6
CAD
CAAAAA17076-22
CAAAA17076-06
CAAAA17076-14
3-Bromo-5-iodobenzoic acid 97%
3-Bromo-5-iodobenzoic acid
Purity: 97%
CAS No.: 188815-32-9
Molecular Formula: C7H4BrIO2
Formula Weight: 326.92
Melting Point: 218-221°
Beilstein Reference: 3240837
Formula:
C₇H₄BrIO₂ MW: 326.92 g/mol Melting Pt: 218…221 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191851 CAS Number: 188815-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...