2,2-Dimethylbutyric acid 97%

Supplier: Thermo Scientific Chemicals
Warning

A17072-09 A17072-18 A17072-30
CAAAA17072-09EA 42.66 CAD
CAAAA17072-09 CAAAA17072-18 CAAAA17072-30
2,2-Dimethylbutyric acid 97%
2,2-Dimethylbutyric acid

Purity: 97% CAS No.: 595-37-9 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 90°(194°F) Refractive Index: 1.4154 Odor: Pungent Melting Point: -14° Boiling Point: 186° Beilstein Reference: 1720666 UN No.: 3265 MDL No.: MFCD00004200

Formula: C₆H₁₂O₂
Boiling Pt: 186 °C
Melting Pt: –14 °C
Density: 0.926
Flash Pt: 90 °C(194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004200
CAS Number: 595-37-9
EINECS: 209-865-0
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR