2-Methylimidazole 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17063-0E
A17063-22
A17063-36
CAAAA17063-0EEA
597.24
CAD
CAAAA17063-0E
CAAAA17063-22
CAAAA17063-36
2-Methylimidazole 97%
2-Methylimidazole
Purity: 97%
CAS No.: 693-98-1
Molecular Formula: C4H6N2
Formula Weight: 82.11
Flash Point: 155°(311°F)
Melting Point: 142-144°
Boiling Point: 267-268°
Beilstein Reference: 1368
UN No.: 3259
MDL No.: MFCD00005190
Formula:
C₄H₆N₂ Boiling Pt: 267…268 °C Melting Pt: 142…144 °C Density: 0.99 g/cm³ (25 °C) Flash Pt: 155 °C (311 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005190 CAS Number: 693-98-1 EINECS: 211-765-7 UN: 3263 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...