2,4-Difluorobenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17052-06
A17052-14
CAAAA17052-06EA
94.48
CAD
CAAAA17052-06
CAAAA17052-14
2,4-Difluorobenzaldehyde 98%
2,4-Difluorobenzaldehyde
Purity: 98%
CAS No.: 1550-35-2
Molecular Formula: C7H4F2O
Formula Weight: 142.11
Flash Point: 55°(131°F)
Refractive Index: 1.4980
Melting Point: 2-3°
Boiling Point: 65-66°/17mm
Beilstein Reference: 2243422
UN No.: 1989
MDL No.: MFCD00010326
Formula:
C₇H₄F₂O Boiling Pt: 65…66 °C/17 mm Melting Pt: 2…3 °C Density: 1.299 Flash Pt: 55 °C (131 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010326 CAS Number: 1550-35-2 EINECS: 216-287-2 UN: 1989 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...