DL-α-Tocopherol (Vitamin E) 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17039-0B
A17039-18
A17039-30
CAAAAA17039-0BEA
1235.56
CAD
CAAAAA17039-0B
CAAAA17039-18
CAAAA17039-30
DL-α-Tocopherol (Vitamin E) 97+%
DL-α-Tocopherol
Purity: 97+%
CAS No.: 10191-41-0
Molecular Formula: C29H50O2
Formula Weight: 430.72
Flash Point: 240°(464°F)
Physical Form: Viscous Liquid
Density: 0.950
Refractive Index: 1.5050
Sensitivity: Light Sensitive
Melting Point: 2-4°
Boiling Point: 200-220°/0.1mm
Beilstein Reference: 94012
Merck Reference: 14,10021
MDL No.: MFCD00072051
Formula:
C₂₉H₅₀O₂ Boiling Pt: 200…220 °C/0.1 mm Melting Pt: 2…4 °C Density: 0.950 Flash Pt: 240 °C(464 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00072051 CAS Number: 10191-41-0 EINECS: 233-466-0 Merck Index: 14,10021 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...