CAPS (3-(cyclohexylamino)propanesulfonic acid) 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17037-36
A17037-14
A17037-22
CAAAAA17037-36EA
437.66
CAD
CAAAAA17037-36
CAAAA17037-14
CAAAA17037-22
CAPS (3-(cyclohexylamino)propanesulfonic acid) 99%
CAPS (3-(Cyclohexylamino)propanesulphonic acid)
Purity: 99%
CAS No.: 1135-40-6
Molecular Formula: C9H19NO3S
Formula Weight: 221.32
Odor: Odorless
Melting Point: 324°
Beilstein Reference: 2835588
Merck Reference: 14,1767
MDL No.: MFCD00003837
Formula:
C₆H₁₁NH(CH₂)₃SO₃H Melting Pt: 324 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003837 CAS Number: 1135-40-6 EINECS: 214-492-1 Merck Index: 14,1767 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...