Quinine hemisulfate monohydrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Quinine sulphate, Quinine sulfate, Quinine sulfate (2:1) (salt) dihydrate, Quinine sulphate (2:1) (salt) dihydrate
A17036-09
A17036-18
A17036-30
CAAAA17036-09EA
74.1
CAD
CAAAA17036-09
CAAAA17036-18
CAAAA17036-30
Quinine hemisulfate monohydrate 99%
Quinine hemisulfate monohydrate
A potassium channel blocker
Purity: 99%
CAS No.: 6119-70-6
Molecular Formula: C20H24N2O2.0.5H2SO4.H2O
Formula Weight: 391.47 (373.46anhy)
Odor: Odorless
Melting Point: 217-218° dec.
Beilstein Reference: 6113937
Merck Reference: 14,8061
Optical Rotation: -241° (c=2 in 0.1M HCl)
MDL No.: MFCD00150792
Formula:
C₂₀H₂₄N₂O₂·0.5H₂SO₄·H₂O MW: 391.47 g/mol (373.46 anhy) Melting Pt: 217…218 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00150792 CAS Number: 6119-70-6 EINECS: 212-359-2 Merck Index: 14,8061 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...