1-Bromo-2-iodobenzene 98+% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromo-1-iodobenzene, 2-Bromoiodobenzene

Stabilization: Stabilised with copper

A17020-06 A17020-14 A17020-22
CAAAA17020-06EA 74.42 CAD
CAAAA17020-06 CAAAA17020-14 CAAAA17020-22
1-Bromo-2-iodobenzene 98+% stabilized
1-Bromo-2-iodobenzene

Purity: 98+% CAS No.: 583-55-1 Molecular Formula: C6H4BrI Formula Weight: 282.91 Flash Point: 110°(230°F) Refractive Index: 1.6645 Melting Point: 9-10° Boiling Point: 257° Beilstein Reference: 1855309 MDL No.: MFCD00001030

Formula: C₆H₄BrI
MW: 282.91 g/mol
Boiling Pt: 257 °C
Melting Pt: 9…10 °C
Density: 2.256
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001030
CAS Number: 583-55-1
EINECS: 209-508-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR